Orders LinkContactUsLinkComponent
BioPharm Plus Platinum-Cured Silicone Tubing | Avantor
BioPharm Plus Platinum-Cured Silicone Tubing
Advantages:
  • Up to three times the working life of other platinum silicone tubings
  • Excellent biocompatibility; no leachable additives, DOP, or plasticizers; odorless and nontoxic, fungus resistant; no taste imparted to transported fluids.
  • Ultra-smooth inner surface minimizes particle entrapment.
  • Ideal for laboratory, food/beverage, and pharmaceutical applications.
  • Weather, ozone, corona, and radiation resistant.
Limitations:
  • Do not use with concentrated solvents, oils, or acids.
  • Relatively high gas permeability.
Application Suitability:
Acids:Poor
Alkalies:Poor
Organic solvents:Not recommended
Pressure:Fair
Vacuum:Good
Viscous fluids:Fair
Sterile fluids:Excellent
Physical Characteristics and Composition:
  • Thermal set rubber.
  • Siloxane polymers and amorphous silica.
  • Excellent compression strength.
  • Soft material.
  • Translucent, clear to light amber.
Temperature Range:
-60 to 460°F (-51 to 232°C)
Meets Classifications:
  • USP Class VI and 3A sanitary standards
  • FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
CO2:25,147
H2:-
O2:4715
N2:2284
Cleaning/Sterilization:
  • Sterilize by ethylene oxide (ETO), autoclave, or gamma irradiation