Orders LinkContactUsLinkComponent
UV/VIS Spectroscopy | Avantor
UV/VIS Spectroscopy
UV/Vis analysis with a spectrophotometer can require different types of chemicals. First, a specific grade of solvents is developed to avoid optical impurities of your sample. Second, the spectrophotometer needs to be calibrated using UV/Vis standard kits. Third, color reference solution kits can save you time in preparing the reference solutions yourself.
Frequently Asked Questions
What kind of standards are available to calibrate my UV/Visible spectrophotometer?
Different standards in ampoules delivered in kits with a complete Certificate of Analysis are available to check parameters such as:
  • Absorbance control
  • Stray light testing
  • Resolution power testing
  • Wavelength testing
Literature