Orders LinkContactUsLinkComponent
Metal trace analysis (AAS/ICP) | Avantor
Metal Trace Analysis (AAS/ICP)
In metal analysis, elemental evidence is now gathered from the tiniest traces in the ppm, ppb and ppt ranges with sophisticated instrumentation like AAS, ICP-OES or, more recently, ICP-MS.
To analyze samples properly, they need a suitable pre-treatment. A sample must first be dissolved, digested or fused. All the auxiliary reagents used must perform these tasks, but be invisible in the final analysis.
Avantor®, J.T.Baker® and Merck standards and reagents are specifically produced to give you that blank background so that you can solve even the most baffling cases.
  • Chemicals for sample preparation for trace analysis
  • Standards for calibration
  • Equipment for trace analysis
  • Related documents for metal trace detection
A complete offering with maximum permitted levels of contaminants in ppb and ppt for ultra-trace analysis.
Highly pure acids with maximum permitted levels of contaminants in ppm and ppt for ultra-trace analysis.
Complete range of AAS standards of 100 mg/L available in different packs, traceable to NIST, and delivered with Certificate of Analysis (CoA).
Full range of standards at 10, 1,000 and 10,000 mg/I.
Multi-element solutions for perfect equipment calibration. Traceable to NIST and delivered with a Certificate of Analysis (CoA).
A variety of matrix modifiers for GFAA, CRM, VHG, Promochem and more.
A wide assortment of acids, caustics and salts for many inorganic and analytical applications. 99 percent stock availability.
VWR and Merck alkalis are available for many inorganic and analytical applications.
Manufactured under strictly-controlled conditions.
Ideal for sample preparations. Manufactured under strictly-controlled conditions. Traceable to NIST and delivered with a certificate of a analysis (CoA).
What is Atomic Spectroscopy?
Atomic spectroscopy lets analysts determine the composition of liquid or solid samples by measuring the amount of light absorbed by atoms and ions in the material at specific wavelengths.
By applying electromagnetic radiation to a compound, it puts atoms in the sample into an excited state. As they return to their original condition, they emit energy as light. By measuring the wavelength and intensity of the emitted light, analysts can identify the atoms in the sample.