Orders LinkContactUsLinkComponent
metteldo_1920x760.jpg
Conductivity Measurement
Conductivity measurement of an aqueous solution is a method used to determine amount of nutrients, salts and impurities. Conductivity standards are crucial to calibrate your conductivity meter. Depending on you measurement range, different standards are available. You can buy them in bottles or practical sachets to ensure a 'fresh' solution every time you calibrate.
Conductivity measurement​ | Avantor
Product categories
A very large range with different concentrations from 15 to 100,000 microSiemens/cm for analyzing samples or calibrating conductivity cells. Produced according to ISO 17025 / ISO 17034
Conductivity standards in bottles
A very large range with different concentrations from 15 to 100,000 microSiemens/cm for analysing samples or calibrating conductivity cells. Produced by a supplier following ISO 17025 and Guide 34
Frequently Asked Questions
What are the advantages of using conductivity standards in sachets?
There is nothing to mix or measure. Just open one sachet and your fresh solution is ready to use. Our sachets are perfect for the field and lab since no extra glassware or measuring devices are needed, and nothing needs to be washed. These single-dose sachets ensure that you have just the right amount of solution every time, preventing cross-contamination of your samples.
  • Pocket size
  • Freshness sealed
  • Reliable, easy to use and accurate
  • Traceability to NIST
  • Batch-specific Certificates of Analysis included in each pack showing
  • Measurement uncertainty
  • pH values at various T°
  • Expiry dates
  • Traceability to NIST
How are conductivity and TDS related?
Salts, minerals and dissolved gases contribute to conductivity, meaning conductivity indicates the amount of dissolved materials in a solution. TDS can give you a fairly accurate measurement when comparing the status of a single source (such as NaCl), yet when comparing two different types of solutions it may be error-prone. Calibrate the meter using the same dissolved materials that are in the test solution.
How does temperature affect conductivity readings?
It depends on the solution you are measuring. The effect of temperature is greatest in low conductivity (or low ionic strength) solutions. In most cases, there will be a 2% increase per °C for most aqueous solutions. However, organics have very different temperature curves.
What is the difference between conductivity and salinity?
The probe is the same for both, but a salinity meter has a correction factor applied to the reading. The correction factor takes the conductivity reading and converts it to ppm of a specific salt (which varies by manufacturer).
Are conductivity probes interchangeable with meters?
No. The cell constant may be different and pin configurations are typically different. The type of thermistor used for temperature compensation is also different.
How and when do I need to calibrate the probe?
Use a standard solution in the range of samples you are testing to calibrate the probe. Place the probe in the standard solution, condition and rinse the probe in a second sample of the standard solution. Then use a third sample of standard solution to calibrate and adjust the cell constant until the specified value is displayed. Recalibrate when you change ranges, or if readings seem inaccurate.
Literature