$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
QUALITATIVE STEC IN EDIBLE PK5
SXCMPT-029B
:
SPEX CERTIPREP LLC
New Product
QUALITATIVE STEC IN EDIBLE PK5
SXCMPT-029B
:
SPEX CERTIPREP LLC
:
CMPT-029B
New Product
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International