N-METHYL-N-PERFLUOROBUTANESULFONYLGL 1ML
Catalog # SX14172.7-50ME
Supplier: SPEX CERTIPREP LLC
New Product
N-METHYL-N-PERFLUOROBUTANESULFONYLGL 1ML
Catalog # SX14172.7-50ME
Supplier: SPEX CERTIPREP LLC
Supplier Number: 14172.7-50-ME
New Product
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Difco™ Granulated Agar, BD Biosciences
Supplier: BD
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Single-Use Needles, BD Medical
Supplier: BD
Corning® Trypsin
Supplier: Corning
Corning® Cell Recovery Solution, Corning
Supplier: Corning
Solvent Resistant Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Needles for BD Eclipse™ Syringes, BD
Supplier: BD
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
ELISA KIT HCG < 5 MIU/ML 96 WELLS 25 UL
Supplier: DRG International
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Canada
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach