TOX IN SOIL PT EXPRESS
Catalog # SXSQCO-038B
Supplier: SPEX CERTIPREP LLC
New Product
TOX IN SOIL PT EXPRESS
Catalog # SXSQCO-038B
Supplier: SPEX CERTIPREP LLC
Supplier Number: SQCO-038B
New Product
Frequently Bought Together
InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Supplier: Mettler Toledo
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Calcein AM fluorescent dye, Corning®
Supplier: Corning
INDICATOR PAPER WATOR DARK BLUE 78X40 MM
Supplier: Macherey-Nagel
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
Free and Total Chlorine Test Strips, Hach
Supplier: Hach
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Iodine (iodine-iodide) 0.100 N (N/10)
Supplier: Ricca Chemical
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
Reagent Sets for Colorimeters, Hach
Supplier: Hach
CASSETTE BIOPSY BULK LILAC CS1500
Supplier: Simport Scientific
High Sensitivity DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Succinimide 98+%
Supplier: Thermo Scientific Chemicals