TPH IN SOIL PT EXPRESS
Catalog # SXSQCI-140B
Supplier: SPEX CERTIPREP LLC
New Product
TPH IN SOIL PT EXPRESS
Catalog # SXSQCI-140B
Supplier: SPEX CERTIPREP LLC
Supplier Number: SQCI-140B
New Product
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Quality Economy Brand Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
Leakbuster™ Specimen Containers
Supplier: Starplex
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
NEEDLE 20GA 1IN SS PINKPK50
Supplier: Loctite
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Quinoline 98%
Supplier: Thermo Scientific Chemicals
Calcein AM fluorescent dye, Corning®
Supplier: Corning
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Buffer Capsules, Oakton® Instruments
Supplier: Antyila Scientific
Silicone emulsion, Antifoam B®, Laboratory Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
TelN Protelomerase
Supplier: New England Biolabs (NEB)
Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
tert-Butyl methyl ether ≥99% ACS, VWR Chemicals BDH®
Supplier: VWR International