HEXAVALENT CHROMIUM PT EXPRESS
Catalog # SXSQCI-003B
Supplier: SPEX CERTIPREP LLC
New Product
HEXAVALENT CHROMIUM PT EXPRESS
Catalog # SXSQCI-003B
Supplier: SPEX CERTIPREP LLC
Supplier Number: SQCI-003B
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Ammonium persulfate (APS) ≥98.0% ACS analytical reagent
Supplier: VWR International
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Phenols, CHEMets Visual Kit, CHEMetrics
Supplier: CHEMetrics
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Sodium hydroxide ≥99% (NaOH basis by acidimetry), pellets, EMSURE® ISO for analysis, Supelco®
Supplier: MilliporeSigma
Malonic Acid
Supplier: Ward's Science
Nitrite, CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: CHEMetrics