MEAT HOMOGENATE 50G PT EXPRESS PK2
Catalog # SXFCPT-005B
Supplier: SPEX CERTIPREP LLC
New Product
MEAT HOMOGENATE 50G PT EXPRESS PK2
Catalog # SXFCPT-005B
Supplier: SPEX CERTIPREP LLC
Supplier Number: FCPT-005B
New Product
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Counting Chambers, C-Chip, NanoEntek
Supplier: NANOENTEK USA INC.
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents, Thermo Scientific
Supplier: Thermo Fisher Scientific
Hazmat Boot Covers, Onguard
Supplier: Onguard Industries
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Sol-M® Standard Insulin Syringes
Supplier: SOL-MILLENNIUM CANADA, INC. CA
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
Chloroform ≥99.8% (chloroform + ethanol basis) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
Potassium antimonate trihydrate ≥94%
Supplier: Thermo Scientific Chemicals
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Plastic Dressing Forceps, Disposable Grade, Sklar
Supplier: Sklar
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC