MBAS WASTEWATER PT EXPRESS PK2
Catalog # SXQCI-124B
Supplier: SPEX CERTIPREP LLC
New Product
MBAS WASTEWATER PT EXPRESS PK2
Catalog # SXQCI-124B
Supplier: SPEX CERTIPREP LLC
Supplier Number: QCI-124B
New Product
Frequently Bought Together
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
VACUETTE® Specimen Collection System, Greiner Bio-One
Supplier: Greiner Bio-One
Diethyl ether ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Supplier: Ricca Chemical
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Supplier: Cambridge Diagnostic Products
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
Sodium molybdate(VI) dihydrate 99.5-103.0% ACS
Supplier: Thermo Scientific Chemicals
EZ Double Cytofunnel™, Epredia
Supplier: Epredia
L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
BRUSH CYTOLOGY STERILE 100/PK
Supplier: Medical Packages
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
Dithiothreitol (DTT, Cleland's reagent) ≥98% (by iodometry)
Supplier: Enzo Life Sciences