ELISA KIT HUMAN IL-28B A2216 48TEST
Catalog # 78008-268
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT HUMAN IL-28B A2216 48TEST
Catalog # 78008-268
Supplier: ANTIBODIES.COM LLC
Supplier Number: A2216-48
New Product
Frequently Bought Together
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Sol-M® Standard Syringes without Needle
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
DPD Total Chlorine Reagent Powder Pillows, 5 ml, Hach
Supplier: Hach
PEI MAX® Transfection Reagents (Linear Polyethylenimine Hydrochloride)
Supplier: KYFORA BIO LLC.
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Annexin V Binding Buffer, BioLegend®
Supplier: Biolegend
Human Recombinant FGF-basic (from E. coli)
Supplier: Peprotech
NEB®10-beta Competent <i>E.coli</i> (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
Formalin prefilled 10%
Supplier: Mortech Manufacturing