ELISA KIT DYNAMIN 2 A2233 48TEST
Catalog # 78008-296
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT DYNAMIN 2 A2233 48TEST
Catalog # 78008-296
Supplier: ANTIBODIES.COM LLC
Supplier Number: A2233-48
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
TEGADERM DRESSING 6X7CM 100'S
Supplier: Safecross First Aid Limited
Water for HPLC, Supelco®
Supplier: MilliporeSigma
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
Corning® Donor Horse Serum, US Sourced, Corning
Supplier: Corning
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Methyl Orange
Supplier: Ward's Science
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Alkalinity Test Kit, Model AL-AP, Hach
Supplier: Hach
Ammonia solution 28,0 - 30,0%, GR ACS, Supelco®
Supplier: MilliporeSigma