APTAMER AFLATOXIN M1 A320963 25UG
Catalog # 78000-539
Supplier: ANTIBODIES.COM LLC
New Product
APTAMER AFLATOXIN M1 A320963 25UG
Catalog # 78000-539
Supplier: ANTIBODIES.COM LLC
Supplier Number: A320963-25
New Product
Frequently Bought Together
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
AccuStart™ II GelTrack PCR SuperMix, QuantaBio
Supplier: Quantabio
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Naphthalene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Isobutyl acetate 98%
Supplier: Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane ≥99%
Supplier: Thermo Scientific Chemicals
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences