ASSAY FREE CHOLESTEROL A319676 96TEST
Catalog # 78000-188
Supplier: ANTIBODIES.COM LLC
New Product
ASSAY FREE CHOLESTEROL A319676 96TEST
Catalog # 78000-188
Supplier: ANTIBODIES.COM LLC
Supplier Number: A319676-96
New Product
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Parasitology Control Slides, Azer Scientific
Supplier: Azer Scientific Inc.
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Tru-Punch™ Sterile Disposable Biopsy Punches, Sklar
Supplier: Sklar
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
SYRINGE 1ML LS W/27X 3/8 NEEDLE IB PK100
Supplier: BD
Single-Use Needles, BD Medical
Supplier: BD
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
Iron(III) chloride hexahydrate 97%
Supplier: Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
LE422 Micro pH Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
COMPLEMENT GUINEA PIG NON-STERILE 10ML
Supplier: Rockland Immunochemical
TelN Protelomerase
Supplier: New England Biolabs (NEB)