ELISA KIT MOUSE IFN GAMMA A270541 96TEST
Catalog # 77998-218
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE IFN GAMMA A270541 96TEST
Catalog # 77998-218
Supplier: ANTIBODIES.COM LLC
Supplier Number: A270541-96
New Product
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Reinforced Clostridial Medium, BD Diagnostics
Supplier: BD
Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Trichloroacetic acid (TCA) ≥99% ACS
Supplier: Thermo Scientific Chemicals
Difco™ Baird-Parker Agar Base, BD
Supplier: BD
Toluene ACS
Supplier: INNOVA CHEMICALS CA
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
Glycerine 99+%
Supplier: Thermo Scientific Chemicals
Adjustable Well Plate Stand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Hydrochloric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC