Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
FLASK FERNBACH DESIGN BAFFLED 5 L CS4
TXLAMG431684
:
AVANTOR FLUID HANDLING, LLC
New Product
FLASK FERNBACH DESIGN BAFFLED 5 L CS4
TXLAMG431684
:
AVANTOR FLUID HANDLING, LLC
:
AMG-431684
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International