LITHIUM ION STANDARD SOLUTION 50ML
Catalog # 77974-296
Supplier: FUJIFILM IRVINE SCIENTIFIC WAKO CHE
New Product
LITHIUM ION STANDARD SOLUTION 50ML
Catalog # 77974-296
Supplier: FUJIFILM IRVINE SCIENTIFIC WAKO CHE
Supplier Number: 126-03391
New Product
Frequently Bought Together
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Leakbuster™ Specimen Containers
Supplier: Starplex
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
Supplier: VWR
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
Supplier: Hach
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
Preserved Starfish
Supplier: Avantor
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
Ionic strength adjustor (ISA) solution for use with sodium ion selective electrode
Supplier: Thermo Scientific Chemicals
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Control Slides, Special Stains, Copper
Supplier: Mortech Manufacturing
Chloroform ≥99.8% (chloroform + ethanol basis) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
Lysing Matrix B, MP Biomedicals
Supplier: MP Biomedicals
ISOPROPYL ALCOHOL 100% 1US GAL CS4
Supplier: CANCER DIAGNOSTICS, INC.