BUFFER SOLUTION STANDARD PH9.18 500ML
Catalog # 77973-919
Supplier: FUJIFILM IRVINE SCIENTIFIC WAKO CHE
New Product
BUFFER SOLUTION STANDARD PH9.18 500ML
Catalog # 77973-919
Supplier: FUJIFILM IRVINE SCIENTIFIC WAKO CHE
Supplier Number: 028-03205
New Product
Frequently Bought Together
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, deionized VWR®
Supplier: VWR
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Histology Ultra Fine-Tip Pen
Supplier: VWR International
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal Health
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach
XLD ( Xylose Lysine Deoxycholate) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Corning® Cell Recovery Solution, Corning
Supplier: Corning
CL17 DPD Indicator, Hach
Supplier: Hach
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences