Adjustable Volume, Single Channel Pipettes
Adjustable volume, single channel pipette, 1 - 10 ml, Gray
Catalog # 77973-376
New Product
Supplier: CELLTREAT Scientific Products
Supplier #: 230314
Adjustable Volume, Single Channel Pipettes
Catalog # 77973-376
Supplier: CELLTREAT Scientific Products
New Product
Jump to:
Product Details & Documents
Adjustable volume pipettes allow users to set and change liquid volumes for precise aspiration and dispensing in labs.
- High precision adjustable pipette with easy-to-read volume display
Continuously adjustable with locking ring to prevent volume changes while pipetting
Lightweight design and easy operation with either hand
Reduced operating forces; plunger is easy to press and dispense
Strong magnets are included on both sides for increased versatility
Pipet sizes, colors, and types can be viewed through the side windows
Side bracket can hold pipet fillers, bulbs, and pumps
Specifications
- Volume:1 - 10 ml
- Description:Adjustable volume, single channel pipette
- Color code:Gray
Accessories
Product Information
- Catalog #AvailabilityPrice
- Plastic storage rack, magnetized, white