Low Retention Pipette Tip Reload System, Non-sterile
300 µl, -, Natural, 960 tips/reload system
Catalog # 77972-336
New Product
Supplier: CELLTREAT Scientific Products
Supplier #: 229055
Low Retention Pipette Tip Reload System, Non-sterile
Catalog # 77972-336
Supplier: CELLTREAT Scientific Products
New Product
Jump to:
Product Details & Documents
Pipette tip reload systems quickly and easily refill pipette tip racks, saving time and reducing plastic waste.
- Save time in refilling empty pipette racks
Secure locking tab keeps reload system closed and prevents tumbling tips
Reduce plastic waste and save time by refilling empty racks
Specifications
- Volume:300 µl
- Tip color:Natural
- Packed:960 tips/reload system
- Sterile:-