Orders LinkContactUsLinkComponent
Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
Bulk packed, sterile, 9"
Catalog # 77972-594
New Product
Supplier #:  229286
Plasteur® Pasteur Pipets
Catalog # 77972-594
New Product
Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
Bulk packed, sterile, 9"
Price
$259.12
Log in for availability
Case of 500
Product Details & Documents
Plasteur pipets for transferring liquids.
  • CELLTREAT® exclusive product
    Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
    Flexible plastic helps prevent glove or skin punctures
    Available packaged sterile individually wrapped or bulk packed
    Available packaged non-sterile in re-sealable bags or tab lock boxes
    Eliminate sharps disposal costs
Specifications
  • Description:
    Bulk packed, sterile
  • Length:
    9"