Plasteur® Pasteur Pipets
Bulk Packed, tab lock box, non-sterile, 5.75"
Catalog # 77972-576
New Product
Supplier: CELLTREAT Scientific Products
Supplier #: 229274
Jump to:
Product Details & Documents
Plasteur pipets for transferring liquids.
- CELLTREAT® exclusive product
Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
Flexible plastic helps prevent glove or skin punctures
Available packaged sterile individually wrapped or bulk packed
Available packaged non-sterile in re-sealable bags or tab lock boxes
Eliminate sharps disposal costs
Documents
Avantor CELLTREAT Polypropylene Pasteur Pipets.pdf
Product Literature
Specifications
- Description:Bulk Packed, tab lock box, non-sterile
- Length:5.75"