Deep Well Storage Plates, Polypropylene
48-well plate, rectangle well, U-bottom, Polypropylene, 4.6 ml, Natural, Non sterile
77972-968
New Product
: 229578
Deep well storage plates help minimize the risk of sample evaporation and contamination in sample storage, liquid handling, and high-throughput screening.
- Square to round transition of V-bottom maximizes working volume
- Pyramid bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than a vortex when agitated
- Round bottom with round wells enables use with magnetic beads
Deep well storage plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene material offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications.
- Sterility:Non sterile
- Volume:4.6 ml
- Color:Natural
- Description:48-well plate, rectangle well, U-bottom
- Material:Polypropylene