RECOMBINANT RAT IL-1RA PR AB322101-10UG
Catalog # ABCA_AB322101-10UG
Supplier: Abcam
New Product
RECOMBINANT RAT IL-1RA PR AB322101-10UG
Catalog # ABCA_AB322101-10UG
Supplier: Abcam
Supplier Number: AB322101-10UG
New Product
Frequently Bought Together
Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Premium Aluminum Foil
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Shelf Liners
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Reynolds Wrap® Aluminum Foil
Supplier: Antyila Scientific
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Hanus solution (Iodine bromine) for iodine absorption number of fats and oils
Supplier: Ricca Chemical
Fluoride Standard Solutions, Traceable to NIST, Hach
Supplier: Hach