RECOMBINANT MONKEY IL-1BE AB320810-10UG
Catalog # ABCA_AB320810-10UG
Supplier: Abcam
New Product
RECOMBINANT MONKEY IL-1BE AB320810-10UG
Catalog # ABCA_AB320810-10UG
Supplier: Abcam
Supplier Number: AB320810-10UG
New Product
Frequently Bought Together
Hydrochloric acid (2.87 - <lt/> 7.8 mol/l; 2.87 - <lt/> 7.8 N) 6.0 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
2-Propanol ≥70%, VWR Chemicals BDH®
Supplier: VWR International
SWAB CLEANER KIT PK50
Supplier: Advanced Instruments
Potassium iodide 20% (w/v) in aqueous solution stabilized
Supplier: Ricca Chemical
Ethanol denatured ACS (denatured with methanol and ethyl acetate)
Supplier: INNOVA CHEMICALS CA