SERINE PROTEASE ASSAY AB270782-100TEST
Catalog # ABCA_AB270782-100T
Supplier: Abcam
New Product
SERINE PROTEASE ASSAY AB270782-100TEST
Catalog # ABCA_AB270782-100T
Supplier: Abcam
Supplier Number: AB270782-100TEST
New Product
Frequently Bought Together
Ward's® Preserved Lubber Grasshopper
Supplier: Ward's Science
Potassium hydrogen phthalate 99.95-100.05% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
INDICATOR PAPER WATOR DARK BLUE 78X40 MM
Supplier: Macherey-Nagel
Acetylacetone 99%
Supplier: Thermo Scientific Chemicals
Orion™ Sure-Flow® pH Electrode 9172BNWP, Thermo Scientific
Supplier: Thermo Fisher Scientific
Paraformaldehyde 4% in PBS
Supplier: Thermo Scientific Chemicals
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Canada
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
SevenExcellence pH/mV Meters, METTLER TOLEDO®
Supplier: Mettler Toledo
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)