SERINE PROTEASE ASSAY K AB270779-25TEST
Catalog # ABCA_AB270779-25TE
Supplier: Abcam
New Product
SERINE PROTEASE ASSAY K AB270779-25TEST
Catalog # ABCA_AB270779-25TE
Supplier: Abcam
Supplier Number: AB270779-25TEST
New Product
Frequently Bought Together
Sodium chloride solution 0.90% (w/v) in water, VWR Chemicals BDH®
Supplier: VWR International
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Sodium molybdate(VI) dihydrate 99.5-103.0% ACS
Supplier: Thermo Scientific Chemicals