SERINE PROTEASE ASSAY AB270778-100TEST
Catalog # ABCA_AB270778-100T
Supplier: Abcam
New Product
SERINE PROTEASE ASSAY AB270778-100TEST
Catalog # ABCA_AB270778-100T
Supplier: Abcam
Supplier Number: AB270778-100TEST
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sodium Bicarbonate
Supplier: Ward's Science