SERINE PROTEASE ASSAY K AB270776-25TEST
Catalog # ABCA_AB270776-25TE
Supplier: Abcam
New Product
SERINE PROTEASE ASSAY K AB270776-25TEST
Catalog # ABCA_AB270776-25TE
Supplier: Abcam
Supplier Number: AB270776-25TEST
New Product
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Single-Use Needles, BD Medical
Supplier: BD
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research
Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: VWR International
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Phusion High-Fidelity DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide