RECOMBINANT RAT IL-17F PRO AB269244-5UG
Catalog # ABCA_AB269244-5UG
Supplier: Abcam
New Product
RECOMBINANT RAT IL-17F PRO AB269244-5UG
Catalog # ABCA_AB269244-5UG
Supplier: Abcam
Supplier Number: AB269244-5UG
New Product
Frequently Bought Together
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Sleeve Stoppers
Supplier: VWR International
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
VWR® Universal Pipette Tips
Supplier: VWR International
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Falcon® 96-Well Cell Culture Plates, Corning
Supplier: Corning
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
SingleQuots® EGM®-2 Endothelial Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
Single-Use Needles, BD Medical
Supplier: BD
Corning® Trypsin
Supplier: Corning