RECOMBINANT RAT IL-17F P AB269244-500UG
Catalog # ABCA_AB269244-500U
Supplier: Abcam
New Product
RECOMBINANT RAT IL-17F P AB269244-500UG
Catalog # ABCA_AB269244-500U
Supplier: Abcam
Supplier Number: AB269244-500UG
New Product
Frequently Bought Together
Sodium chloride solution 0.90% (w/v) in water, VWR Chemicals BDH®
Supplier: VWR International
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
Nunc-Immuno™ MicroWell™ 96-Well Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
Parasitology Control Slides, Azer Scientific
Supplier: Azer Scientific Inc.
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Magnesium chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Potassium iodate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Iron(III) chloride hexahydrate 97%
Supplier: Thermo Scientific Chemicals
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
3-Iodo-L-tyrosine 98%
Supplier: Thermo Scientific Chemicals
TelN Protelomerase
Supplier: New England Biolabs (NEB)
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech