RECOMBINANT MOUSE IL-17F AB269190-500UG
Catalog # ABCA_AB269190-500U
Supplier: Abcam
New Product
RECOMBINANT MOUSE IL-17F AB269190-500UG
Catalog # ABCA_AB269190-500U
Supplier: Abcam
Supplier Number: AB269190-500UG
New Product
Frequently Bought Together
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
VWR® Reagent alcohol
Supplier: VWR International
Sol-M® Standard Syringes without Needle
Supplier: SOL-MILLENNIUM CANADA, INC. CA
E 5 LIQ MACH DISHWSH UN 4L
Supplier: Ostrem Chemical
KIMBLE® Safety Tip Needles with Blunt Ends, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Glycerine 99+%
Supplier: Thermo Scientific Chemicals
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Chloroform ≥99.9% stabilized, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Hazmat Boot Covers, Onguard
Supplier: Onguard Industries
Pierce™ Immobilised TCEP Disulfide Reducing Gel, Thermo Scientific
Supplier: Thermo Scientific
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Potassium pyroantimonate USP Test Solution (TS)
Supplier: Ricca Chemical
T4 DNA Ligase, New England Biolabs
Supplier: New England Biolabs (NEB)
Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC