Cell Lysis Buffer, Abcam
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This product is manufactured by BioVision, an Abcam company and was previously called 1067 Cell Lysis Buffer.
- Constituents: 1% Alcohol Ethoxylate, 0.88% Sodium chloride, 0.32% Tris HCl
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:Cell Lysis Buffer
- Size:400 ml
- Cat. No.:ABCA_AB288311-400M
- Storage temperature:Store at +4°C.
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



