HUMAN IL-17F ELISPOT AB281968-1X96TESTS
ABCA_AB281968-1X96
: Abcam
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
: VWR International
Hose Clamps, Adjustable, Stainless Steel
: VWR International
Accessories for Micro-Osmometer, Advanced Instruments
: Advanced Instruments
THREADLOCKER 222 BOTTLE 50ML
: Loctite
Phenols, CHEMets Visual Kit, CHEMetrics
: CHEMetrics
Preserved Starfish
: Avantor
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
: VWR International
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
Pierce™ Immobilised TCEP Disulfide Reducing Gel, Thermo Scientific
: Thermo Scientific
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
: Omega Bio-Tek
Acetic Acid
: Ward's Science
1,1'-Ferrocenedicarboxylic acid 98%
: Thermo Scientific Chemicals
2-Deoxy-D-glucose 98%
: Thermo Scientific Chemicals
Dirhenium heptaoxide ≥99.9% (metals basis)
: Thermo Scientific Chemicals