D-SERINE, ENDOGENOUS NMDA R AB120048-1G
Catalog # ABCA_AB120048-1G
Supplier: Abcam
New Product
D-SERINE, ENDOGENOUS NMDA R AB120048-1G
Catalog # ABCA_AB120048-1G
Supplier: Abcam
Supplier Number: AB120048-1G
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Leakbuster™ Specimen Containers
Supplier: Starplex
pH Test Papers, Precision Laboratories
Supplier: Precision Laboratories
Calcium chloride 2.75% (w/v) in aqueous solution
Supplier: Ricca Chemical
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
Supplier: VWR
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Diethyl ether ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Masterflex® L/S® High-Performance Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Tergazyme® Enzyme-Active Powder Detergents
Supplier: Alconox
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
XbaI
Supplier: New England Biolabs (NEB)
INDICATOR PAPER WATOR DARK BLUE 78X40 MM
Supplier: Macherey-Nagel
Dieffenbach Bulldog Clamp, OR Grade, Sklar
Supplier: Sklar