RECOMBINANT HUMAN SERINE AB116196-10UG
ABCA_AB116196-10UG
: Abcam
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
: MilliporeSigma
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
2-Propanol ≥99.5% (by GC), GR ACS, Supelco®
: MilliporeSigma
VWR® Disposable Petri Dishes
: VWR International
Methanol ≥99.90%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Research Chemicals
Cetrimide Select Agar, Hardy Diagnostics
: HARDY DIAGNOSTICS CA
Toluene ACS
: INNOVA CHEMICALS CA