RECOMBINANT HUMAN PML PRO AB112341-10UG
Catalog # ABCA_AB112341-10UG
Supplier: Abcam
New Product
RECOMBINANT HUMAN PML PRO AB112341-10UG
Catalog # ABCA_AB112341-10UG
Supplier: Abcam
Supplier Number: AB112341-10UG
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
2-Propanol ≥99.0% USP, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
L(-)-Proline 99%
Supplier: Thermo Scientific Chemicals
E.Z.N.A.® RNA Isolation Kits, Omega Bio-Tek
Supplier: Omega Bio-Tek
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Supplier: Thermo Scientific Chemicals
Rhizopus stolonifer (nigricans) (-)
Supplier: AMERICAN TYPE CULTURE COLL.
Single-Use Needles, BD Medical
Supplier: BD
Male to Female Extension Lines, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
Vinyl Powder-Free Disposable Gloves, Tronex®
Supplier: Tronex
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris