Human IL-17F ELISPOT Kit (with un-coated plates)
Catalog # ABCA_AB105242-5X96
Supplier: Abcam
New Product
CAS Number:
Human IL-17F ELISPOT Kit (with un-coated plates)
Catalog # ABCA_AB105242-5X96
Supplier: Abcam
Supplier Number: AB105242-5X96TESTS
New Product
CAS Number:
About this item
Frequently Bought Together
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
XLD ( Xylose Lysine Deoxycholate) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Human Recombinant PDGF-AA (from <i>E. coli</i>)
Supplier: Peprotech
SYRINGE WITH LUER SLIP TIP STERILE 10ML
Supplier: BD
Parafilm M, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
1,4-Diacetylbenzene ≥99.0%
Supplier: TCI America
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics