ELISA KIT RAT IL-17F AB239423-1X96TESTS
Catalog # ABCA_AB239423-1X96
Supplier: Abcam
New Product
ELISA KIT RAT IL-17F AB239423-1X96TESTS
Catalog # ABCA_AB239423-1X96
Supplier: Abcam
Supplier Number: AB239423-1X96TESTS
New Product
Frequently Bought Together
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sodium hydroxide, pellets ACS
Supplier: VWR
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
Supplier: VWR
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Bradford's reagent for dye, ready-to-use
Supplier: Thermo Scientific Chemicals
Nitric acid 67 - 70%, ULTREX® II Ultrapure Reagent, Ultrapure for ICP, for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Copper(II) acetate, anhydrous
Supplier: Thermo Scientific Chemicals
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
Scalpel Handles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Amino Acid F Reagent Powder Pillows, 10 ml, Hach
Supplier: Hach
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA