RECOMBINANT MOUSE IL-17F P AB207953-1MG
Catalog # ABCA_AB207953-1MG
Supplier: Abcam
New Product
RECOMBINANT MOUSE IL-17F P AB207953-1MG
Catalog # ABCA_AB207953-1MG
Supplier: Abcam
Supplier Number: AB207953-1MG
New Product
Frequently Bought Together
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
LaMotte® InstaTest® Water Test Strips
Supplier: LaMotte
VACUETTE® Specimen Collection System, Greiner Bio-One
Supplier: Greiner Bio-One
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Eveready® Alkaline Batteries, Bulbtronics
Supplier: Bulbtronics
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
Trimethyl citrate ≥98.0%
Supplier: TCI America
Dieffenbach Bulldog Clamp, OR Grade, Sklar
Supplier: Sklar
4-Methylpiperidine 98+%
Supplier: Thermo Scientific Chemicals
Wood Handle Metal Brush, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Dithiothreitol (DTT, Cleland's reagent) ≥98% (by iodometry)
Supplier: Enzo Life Sciences