RECOMBINANT HUMAN PTS/PTP AB92928-500UG
Catalog # ABCA_AB92928-500UG
Supplier: Abcam
New Product
RECOMBINANT HUMAN PTS/PTP AB92928-500UG
Catalog # ABCA_AB92928-500UG
Supplier: Abcam
Supplier Number: AB92928-500UG
New Product
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Corning® RPMI 1640, Corning
Supplier: Corning
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Water for HPLC, Supelco®
Supplier: MilliporeSigma
Sternheimer Malbin urine sediment stain
Supplier: Globe Scientific
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences