AZASERINE, SERINE DERIVAT AB145895-50MG
Catalog # ABCA_AB145895-50MG
Supplier: Abcam
New Product
AZASERINE, SERINE DERIVAT AB145895-50MG
Catalog # ABCA_AB145895-50MG
Supplier: Abcam
Supplier Number: AB145895-50MG
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
NEEDLE 20GA 1IN SS PINKPK50
Supplier: Loctite
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
VeriSpec™ Iron (Fe) Standards for AAS, RICCA Chemical Company
Supplier: Ricca Chemical
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 97%
Supplier: Thermo Scientific Chemicals
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Buffer, TBS Solution 10X Concentrate (TRIS Buffered Saline), G-Biosciences
Supplier: G-Biosciences
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Supplier: New England Biolabs (NEB)
1-Pentanesulfonic acid sodium salt monohydrate for HPLC
Supplier: Thermo Scientific Chemicals
MinION™ Mk1B DNA/RNA Sequencing Device, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma