MYRIOCIN (ISP-I), SERINE P AB143526-5MG
Catalog # ABCA_AB143526-5MG
Supplier: Abcam
New Product
MYRIOCIN (ISP-I), SERINE P AB143526-5MG
Catalog # ABCA_AB143526-5MG
Supplier: Abcam
Supplier Number: AB143526-5MG
New Product
Frequently Bought Together
NEEDLE 20GA 1IN SS PINKPK50
Supplier: Loctite
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
Hydrogen peroxide 6% in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
BBL™ Mannitol Salt Agar, BD
Supplier: BD
Pancreatin
Supplier: Ward's Science
Exel International® Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Caffeine ≥99.7%
Supplier: Thermo Scientific Chemicals
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
Gel Loading Dye Blue (6X), New England Biolabs
Supplier: New England Biolabs (NEB)
TruLine pH 15 Electrode, YSI
Supplier: YSI
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
GASTIGHT® 1000 Series Syringes, Hamilton
Supplier: Hamilton
Norcamphor 98+%
Supplier: Thermo Scientific Chemicals
Anti-TAP1 Rabbit Polyclonal Antibody
Supplier: Proteintech
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC