PKSI-527, PLASMA KALLIKRE AB144543-25MG
Catalog # ABCA_AB144543-25MG
Supplier: Abcam
New Product
PKSI-527, PLASMA KALLIKRE AB144543-25MG
Catalog # ABCA_AB144543-25MG
Supplier: Abcam
Supplier Number: AB144543-25MG
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
Leakbuster™ Specimen Containers
Supplier: Starplex
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
VWR® Sterile 70% Isopropanol
Supplier: VWR International
DNA Clean and Concentrator™ Kits, Zymo Research
Supplier: Zymo Research
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Sodium Bicarbonate
Supplier: Ward's Science
Toluene ACS
Supplier: INNOVA CHEMICALS CA