RECOMBINANT IL-1RA PROTEIN AB229599-1MG
Catalog # ABCA_AB229599-1MG
Supplier: Abcam
New Product
RECOMBINANT IL-1RA PROTEIN AB229599-1MG
Catalog # ABCA_AB229599-1MG
Supplier: Abcam
Supplier Number: AB229599-1MG
New Product
Frequently Bought Together
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
IRRIGATION WATER 500ML
Supplier: Safecross First Aid Limited
Coagulase
Supplier: BD
Single-Use Needles, BD Medical
Supplier: BD
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
BBL™ Phenol Red Broth Base, BD Biosciences
Supplier: BD
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC