RECOMBINANT SERINE/T AB188459-20000UNIT
Catalog # ABCA_AB188459-2000
Supplier: Abcam
New Product
RECOMBINANT SERINE/T AB188459-20000UNIT
Catalog # ABCA_AB188459-2000
Supplier: Abcam
Supplier Number: AB188459-20000UNIT
New Product
Frequently Bought Together
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Mueller Hinton Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Human Recombinant PDGF-AA (from <i>E. coli</i>)
Supplier: Peprotech
Difco™ Baird-Parker Agar Base, BD
Supplier: BD
Phosphate Buffered Saline
Supplier: Ward's Science
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC