Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant Serine/threonine-protein kinase 4/MST-1 (from Baculovirus (Sf9 Insect cells))

Human Recombinant Serine/threonine-protein kinase 4/MST-1 (from Baculovirus (Sf9 Insect cells))

10 µg
Catalog # ABCA_AB60337-10UG
New Product
Supplier:   Abcam
Supplier #:  AB60337-10UG
Human Recombinant Serine/threonine-protein kinase 4/MST-1 (from Baculovirus (Sf9 Insect cells))
Catalog # ABCA_AB60337-10UG
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant Serine/threonine-protein kinase 4/MST-1 (from Baculovirus (Sf9 Insect cells))
Human Recombinant Serine/threonine-protein kinase 4/MST-1 (from Baculovirus (Sf9 Insect cells))

10 µg

CAS Number:  
Price
$944.00
Log in for availability
Each (10µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recombinant human Serine/threonine-protein kinase 4/MST-1 is a Human Full Length protein, expressed in Baculovirus infected Sf9, with >85% purity and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Size:
    10 µg
  • Form:
    Liquid
  • Protein/peptide name:
    Serine/threonine-protein kinase 4/MST-1
  • Protein synonyms:
    KRS2|MST1|STK4|Serine/threonine-protein kinase 4|Mammalian STE20-like protein kinase 1|STE20-like kinase MST1|Serine/threonine-protein kinase Krs-2|MST-1
  • Purity:
    >85%
  • Concentration:
    0.1 mg/ml
  • Source:
    Baculovirus (Sf9 Insect cells)
  • Biological activity:
    Standard
  • Shipping temperature:
    Dry Ice
  • Cat. No.:
    ABCA_AB60337-10UG
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −80 °C
  • Formulation:
    pH: 7.5, Constituents: 25% Glycerol (glycerin, glycerine), 0.87% Sodium chloride, 0.79% Tris HCl, 0.00385% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, 0.0038% EGTA, 0.00292% EDTA, 0.00174% PMSF
  • Conjugation:
    Unconjugated
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.