RECOMBINANT HUMAN SERINE AB152996-10UG
Catalog # ABCA_AB152996-10UG
Supplier: Abcam
New Product
RECOMBINANT HUMAN SERINE AB152996-10UG
Catalog # ABCA_AB152996-10UG
Supplier: Abcam
Supplier Number: AB152996-10UG
New Product
Frequently Bought Together
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
2-Propanol ≥70%, VWR Chemicals BDH®
Supplier: VWR International
Orthophosphoric acid ≥85%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Hose Clamps, Adjustable, Stainless Steel
Supplier: VWR International
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Calcium Nitrate 4-Hydrate
Supplier: Ward's Science
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Supplier: Hardy Diagnostics
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
VeriSpec™ Iron (Fe) Standards for AAS, RICCA Chemical Company
Supplier: Ricca Chemical
SUBSTRATE TMB MEMBRANE PEROXIDASE 1 L
Supplier: Rockland Immunochemical
<i>E. coli</i> Exonuclease III
Supplier: New England Biolabs (NEB)
Ascorbic Acid
Supplier: Ward's Science