ANTIBODY RAT IL-1RA AB253733-10X96TESTS
Catalog # ABCA_AB253733-10X9
Supplier: Abcam
New Product
ANTIBODY RAT IL-1RA AB253733-10X96TESTS
Catalog # ABCA_AB253733-10X9
Supplier: Abcam
Supplier Number: AB253733-10X96TESTS
New Product
Frequently Bought Together
THREADLOCKER 222 BOTTLE 50ML
Supplier: Loctite
Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Supplier: BDH
Leakbuster™ Specimen Containers
Supplier: Starplex
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
American® White Cross Adhesive Bandages, DUKAL™ Corporation
Supplier: Dukal
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Supplier: VWR International
TEST SOLUTION 240 MV 500ML
Supplier: Hanna
Iodine solution 0.1 N, VWR Chemicals BDH®
Supplier: VWR International
VWR® Specimen Containers, PP
Supplier: VWR International
Corning® rat collagen I (from tail tendon)
Supplier: Corning
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International